Product Details

SNP ID
rs13477790
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.4:77633242 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TGATTGACCACATGTGACCGCCTCA[A/C]CATGGACGCCTAAACCTCTTCAATT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
Ptprd PubMed Links

Gene Details

Gene
Ptprd
Gene Name
protein tyrosine phosphatase, receptor type, D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_011211.3 Intron NP_035341.2

View Full Product Details