Product Details
- SNP ID
-
rs3666382
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:34212062 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTTCCTTGTGCATCTTGGACTCCA[C/T]GGTTCTGCATGCAGAGCCCGCTCTT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Kalrn
PubMed Links
Gene Details
- Gene
- Kalrn
- Gene Name
- kalirin, RhoGEF kinase
View Full Product Details