Product Details

SNP ID
rs13481306
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.12:12937225 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AGCTGGAGCGCAGGTCGTTCCGGCG[C/T]TGACGCTCCAGGATGTTGTGGTTGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Mycn PubMed Links

Gene Details

Gene
Mycn
Gene Name
v-myc myelocytomatosis viral related oncogene, neuroblastoma derived (avian)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_008709.3 1464 Silent Mutation NP_032735.3

View Full Product Details