Product Details
- SNP ID
-
rs13482616
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:64815313 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGAATTAAATGCAACTCACATGGGC[C/T]AGCCGGTCGAATCTGGCAAAGAGTT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Adcy8
PubMed Links
Gene Details
- Gene
- Adcy8
- Gene Name
- adenylate cyclase 8
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_009623.2 |
2532 |
Silent Mutation |
|
|
NP_033753.2 |
View Full Product Details