Product Details

SNP ID
rs13475875
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:54744243 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TTAATATCTTTAGAAATGAAGCCAC[A/G]CTTTGTAGGATATCCTAAATCTATG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
4930444A19Rik PubMed Links

Gene Details

Gene
4930444A19Rik
Gene Name
RIKEN cDNA 4930444A19 gene
There are no transcripts associated with this gene.

Gene
Ankrd44
Gene Name
ankyrin repeat domain 44
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081433.3 Intron NP_001074902.2

View Full Product Details