Product Details

SNP ID
rs13475974
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:89750991 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GAGCGCTCTCCTCCGTGACAGGGAC[C/T]GGGGCAATAAGGAGAGCTTGATTAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Agap1 PubMed Links

Gene Details

Gene
Agap1
Gene Name
ArfGAP with GTPase domain, ankyrin repeat and PH domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037136.1 Intron NP_001032213.1
NM_178119.3 Intron NP_835220.1

View Full Product Details