Product Details

SNP ID
rs13476125
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:136117620 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CTCCCATTTCAGCATCTGCTATGAA[A/C]GGGAGTTCTTGGGAGAGGCGGACAT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
Cacna1s PubMed Links

Gene Details

Gene
Cacna1s
Gene Name
calcium channel, voltage-dependent, L type, alpha 1S subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081023.1 5005 Silent Mutation NP_001074492.1
NM_014193.2 5005 Silent Mutation NP_055008.2

View Full Product Details