Product Details

SNP ID
rs6372122
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:51375044 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AGCCACAATTTGGGGGTGACAGAAA[C/T]AAGTAATATATAGGCTGTATAAGAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Col23a1 PubMed Links

Gene Details

Gene
Col23a1
Gene Name
collagen, type XXIII, alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153393.2 Intron NP_700442.2

View Full Product Details