Product Details

SNP ID
rs6280170
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:120479308 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TGGATGAGCGCAGGCGTAGGTGCCC[A/G]CCCTCCGGGGTCCCGCTAATGATTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Hgs PubMed Links

Gene Details

Gene
Hgs
Gene Name
HGF-regulated tyrosine kinase substrate
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159328.1 Intron NP_001152800.1
NM_008244.3 Intron NP_032270.3
Gene
Mrpl12
Gene Name
mitochondrial ribosomal protein L12
There are no transcripts associated with this gene.

View Full Product Details