Product Details

SNP ID
rs6299664
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.14:12025266 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CTCCCACTCCCCACTATCTCTCTAC[A/T]GCTGTTGTTTATAATGAGTTTGGAG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
Ptprg PubMed Links

Gene Details

Gene
Ptprg
Gene Name
protein tyrosine phosphatase, receptor type, G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_008981.3 Intron NP_033007.2

View Full Product Details