Product Details
- SNP ID
-
rs6349691
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:36113613 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGTTTGGTTCACAGAGCCAATCAG[G/T]AAGCAGAGAAAGGGCATGTCAACAT
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Bzw2
PubMed Links
Gene Details
- Gene
- Bzw2
- Gene Name
- basic leucine zipper and W2 domains 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_025840.3 |
|
Intron |
|
|
NP_080116.2 |
View Full Product Details