Product Details

SNP ID
rs13483449
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.18:77637288 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CAGAACTACAGAAAATGGCATCTCC[A/G]TTCAGGCCCGCTGTCCCCCAGGGAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
8030462N17Rik PubMed Links

Gene Details

Gene
8030462N17Rik
Gene Name
RIKEN cDNA 8030462N17 gene
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178670.3 Intron NP_848785.2

View Full Product Details