Product Details

SNP ID
rs29152777
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.12:82873596 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GAGATGCCCTGGACCACCTCATAAC[C/T]GATTTACACACTTCTTTTCATCTCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Rgs6 PubMed Links

Gene Details

Gene
Rgs6
Gene Name
regulator of G-protein signaling 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282061.1 Intron NP_001268990.1
NM_015812.3 Intron NP_056627.1

View Full Product Details