Product Details

SNP ID
rs6225297
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:120139923 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AAGTCCTCCCACACCATGCAAATCC[C/T]ACACATATTCACGAGAAGGATCTCC
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
Slc38a10 PubMed Links

Gene Details

Gene
Slc38a10
Gene Name
solute carrier family 38, member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164798.1 Intron NP_001158270.1
NM_001164799.1 Intron NP_001158271.1
NM_001164800.1 Intron NP_001158272.1
NM_001164801.1 Intron NP_001158273.1
NM_001164802.1 Intron NP_001158274.1
NM_024249.5 Intron NP_077211.4

View Full Product Details