Product Details
- SNP ID
-
rs13481793
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:44884449 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCACAGAAATATTACAGATAAGATT[C/G]TCAAATATTTATCTCCTCCTGGGCG
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Jarid2
PubMed Links
Gene Details
- Gene
- Jarid2
- Gene Name
- jumonji, AT rich interactive domain 2
View Full Product Details