Product Details

SNP ID
rs4179233
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.16:43610765 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AGTTTGTGACAGTGTCCCAGCCCGG[C/T]CTGTCCACCTTTACTGCACAGCTGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Zbtb20 PubMed Links

Gene Details

Gene
Zbtb20
Gene Name
zinc finger and BTB domain containing 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001285805.1 2226 Silent Mutation NP_001272734.1
NM_019778.2 2226 Silent Mutation NP_062752.2
NM_181058.1 2226 Silent Mutation NP_851401.1

View Full Product Details