Product Details

SNP ID
rs3714172
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:75605462 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AGCTGGGATTCTGTTAAGTTTAAGC[T/C]TGGTCTGTGTAAAGACTCTGCTTTA
Phenotype
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
4931413K12Rik PubMed Links

Gene Details

Gene
4931413K12Rik
Gene Name
RIKEN cDNA 4931413K12 gene
There are no transcripts associated with this gene.

Gene
Gm12338
Gene Name
cytochrome c oxidase, subunit VIIc pseudogene
There are no transcripts associated with this gene.

Gene
Pitpna
Gene Name
phosphatidylinositol transfer protein, alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_008850.1 Intron NP_032876.1

View Full Product Details