Product Details
- SNP ID
-
rs6256752
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:21247475 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTGTAAAAGTGATTTATGGACCAG[C/T]TTAAGTGAATTGTTTTCTCTAAGTA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Gsap
PubMed Links
Gene Details
- Gene
- Gsap
- Gene Name
- gamma-secretase activating protein
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_175437.3 |
|
Intron |
|
|
NP_780646.2 |
View Full Product Details