Product Details
- SNP ID
-
rs13481631
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:105643292 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTGAATGAGGTTCATCAGCGCAGC[A/G]CATGAGTCTGAGCATGTTCTGATGT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Atg2b
PubMed Links
Gene Details
- Gene
- Atg2b
- Gene Name
- autophagy related 2B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_029654.4 |
4467 |
Silent Mutation |
|
|
NP_083930.5 |
View Full Product Details