Product Details

SNP ID
rs13476406
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.2:29778860 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GGCCTCCTGGGATTTCAGGTCCAGG[C/T]TGGAGTACCTGGGATTTGGGAGGGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Coq4 PubMed Links

Gene Details

Gene
Coq4
Gene Name
coenzyme Q4 homolog (yeast)
There are no transcripts associated with this gene.

Gene
Trub2
Gene Name
TruB pseudouridine (psi) synthase homolog 2 (E. coli)
There are no transcripts associated with this gene.

View Full Product Details