Product Details

SNP ID
rs13483551
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.19:18886860 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
ACTCACTTAAGGTTCTAACTTTTTC[A/G]TGATGGAAAAGAATCAACAGAAGGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Trpm6 PubMed Links

Gene Details

Gene
Trpm6
Gene Name
transient receptor potential cation channel, subfamily M, member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153417.1 Intron NP_700466.1

View Full Product Details