Product Details
- SNP ID
-
rs13480692
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:88212908 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATTGTAGAAAGTAATCAGGGCACG[C/T]TGTGTCTTGATGTGGTGTGTGTTAG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Ccdc53
PubMed Links
Gene Details
- Gene
- Ccdc53
- Gene Name
- coiled-coil domain containing 53
View Full Product Details