Product Details

SNP ID
rs13482677
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.15:83114395 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TTGACTTCGTAAGCAAGGAGATGTT[C/T]GTGCACTCTGCGAAGAGCCTGCAGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Rrp7a PubMed Links

Gene Details

Gene
Rrp7a
Gene Name
ribosomal RNA processing 7 homolog A (S. cerevisiae)
There are no transcripts associated with this gene.

Gene
Serhl
Gene Name
serine hydrolase-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023475.3 811 Silent Mutation NP_075964.1

View Full Product Details