Product Details

SNP ID
rs13475970
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:88349770 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AGTTTCTCAGACTTCCTGACCACCC[C/T]ATTTCACTGTTAGAATGACGGACCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
4930453O03Rik PubMed Links

Gene Details

Gene
4930453O03Rik
Gene Name
RIKEN cDNA 4930453O03 gene
There are no transcripts associated with this gene.

Gene
Trpm8
Gene Name
transient receptor potential cation channel, subfamily M, member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_134252.3 Intron NP_599013.1

View Full Product Details