Product Details
- SNP ID
-
rs3665690
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:23187039 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTCAGGGGTGCCAACATTACAGAA[A/G]TGAGAAACTATGCTTGATCTAAACT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Eya4
PubMed Links
Gene Details
- Gene
- Eya4
- Gene Name
- eyes absent 4 homolog (Drosophila)
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_010167.4 |
|
Intron |
|
|
NP_034297.2 |
View Full Product Details