Product Details

SNP ID
rs13480824
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.10:128084655 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GAAAACTTGCTACATAGAACCCCCT[A/G]TTACTTTTTTTAAAGCTGTGTTACA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Atp5b PubMed Links

Gene Details

Gene
Atp5b
Gene Name
ATP synthase, H+ transporting mitochondrial F1 complex, beta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016774.3 Intron NP_058054.2
Gene
Baz2a
Gene Name
bromodomain adjacent to zinc finger domain, 2A
There are no transcripts associated with this gene.

Gene
Mir677
Gene Name
microRNA 677
There are no transcripts associated with this gene.

Gene
Ptges3
Gene Name
prostaglandin E synthase 3 (cytosolic)
There are no transcripts associated with this gene.

View Full Product Details