Product Details

SNP ID
rs13481556
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.12:80662553 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CGGTCATCGTCCCGGAAGGAGTGCA[C/T]GCACTTTATGAAGAGGTTCTGGAGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Slc39a9 PubMed Links

Gene Details

Gene
Slc39a9
Gene Name
solute carrier family 39 (zinc transporter), member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_026244.2 846 Silent Mutation NP_080520.2

View Full Product Details