Product Details

SNP ID
rs6206420
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:59220508 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AGAAGCGGAGGACATTGGAATACTC[C/T]GCCATTGTTATCTCTTTGACTGGAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Als2 PubMed Links

Gene Details

Gene
Als2
Gene Name
amyotrophic lateral sclerosis 2 (juvenile)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159948.2 Intron NP_001153420.2
NM_028717.6 Intron NP_082993.4
NM_146109.3 Intron NP_666221.1

View Full Product Details