Product Details

SNP ID
rs13480912
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:23492127 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CATTTCTACAAAAACACATCAACAC[C/T]CTTTCCTTACCTACAACCCAAGTTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Ahsa2 PubMed Links

Gene Details

Gene
Ahsa2
Gene Name
AHA1, activator of heat shock protein ATPase 2
There are no transcripts associated with this gene.

Gene
Usp34
Gene Name
ubiquitin specific peptidase 34
There are no transcripts associated with this gene.

View Full Product Details