Product Details

SNP ID
rs32557211
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:105582090 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AACTCAGCTTGACTGGCCATGGCTG[C/T]TGTGCACATGAGCTCACATTGCTAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Pign PubMed Links

Gene Details

Gene
Pign
Gene Name
phosphatidylinositol glycan anchor biosynthesis, class N
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013784.3 Intron NP_038812.2

View Full Product Details