Product Details

SNP ID
rs13475988
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:93617583 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GAAAGTGAGAATGAATGGTCTGTTC[C/T]TCATTGCTTCACCATCTATGCAGCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Farp2 PubMed Links

Gene Details

Gene
Farp2
Gene Name
FERM, RhoGEF and pleckstrin domain protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145519.2 2571 Missense Mutation NP_663494.2
Gene
Stk25
Gene Name
serine/threonine kinase 25 (yeast)
There are no transcripts associated with this gene.

View Full Product Details