Product Details

SNP ID
rs3164609
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.16:97750659 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AGGGCCATGCCCAGTGGGACTCACT[A/G]TTCAAAAGCCCAACACTGGCACACT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Ripk4 PubMed Links

Gene Details

Gene
Ripk4
Gene Name
receptor-interacting serine-threonine kinase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023663.6 Intron NP_076152.2

View Full Product Details