Product Details

SNP ID
rs6247544
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.2:57315196 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GACGAGGCAGAGCACGGGAGCCCCC[A/G]GGAAACTGGGCAATTGTTTATTGCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Gpd2 PubMed Links

Gene Details

Gene
Gpd2
Gene Name
glycerol phosphate dehydrogenase 2, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145820.1 Intron NP_001139292.1
NM_010274.3 Intron NP_034404.3

View Full Product Details