Product Details
- SNP ID
-
rs13481479
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:58970030 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCCACTAGTGTGTAAACAAAACAA[C/T]AAAATCAACACAAGGATCAGTATGA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Sec23a
PubMed Links
Gene Details
- Gene
- Sec23a
- Gene Name
- SEC23A (S. cerevisiae)
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_009147.2 |
|
Intron |
|
|
NP_033173.2 |
View Full Product Details