Product Details

SNP ID
rs13475968
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:87790830 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AGCAGTCCCCCTTCCTATTTCCAGG[A/G]CTGATTGACTAAAATTAGTGGGGCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Atg16l1 PubMed Links

Gene Details

Gene
Atg16l1
Gene Name
autophagy related 16-like 1 (S. cerevisiae)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001205391.1 Intron NP_001192320.1
NM_001205392.1 Intron NP_001192321.1
NM_029846.4 Intron NP_084122.2
Gene
Scarna6
Gene Name
small Cajal body-specific RNA 6
There are no transcripts associated with this gene.

View Full Product Details