Product Details
- SNP ID
-
rs13476881
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:162071209 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTTCATGCCCCTTTGTACACAATG[A/G]CTGCCTATTCTACTGTCTTTCAAAG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Ptprt
PubMed Links
Gene Details
- Gene
- Ptprt
- Gene Name
- protein tyrosine phosphatase, receptor type, T
There are no transcripts associated with this gene.
View Full Product Details