Product Details

SNP ID
rs13481052
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:60547136 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TCTGGCTGTGTTGATCTCTGCACTT[A/C]AGTGGAGTAGCTTGTAGTGGGAGTA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
Alkbh5 PubMed Links

Gene Details

Gene
Alkbh5
Gene Name
alkB, alkylation repair homolog 5 (E. coli)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172943.4 Intron NP_766531.2

View Full Product Details