Product Details
- SNP ID
-
rs13481456
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:52868858 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTATGAAGAATGGATACTATCAAAA[A/T]AACTTTACCTTTTCTCATATCATGA
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Akap6
PubMed Links
Gene Details
- Gene
- Akap6
- Gene Name
- A kinase (PRKA) anchor protein 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_198111.2 |
|
Intron |
|
|
NP_932779.2 |
View Full Product Details