Product Details
- SNP ID
-
rs13476848
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:153401378 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTATTTGGTCCCGGGAATACAGTT[A/G]CCACCCTTCAGTGCCCCAGGTCTGA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Asxl1
PubMed Links
Gene Details
- Gene
- Asxl1
- Gene Name
- additional sex combs like 1
- Gene
- Nol4l
- Gene Name
- nucleolar protein 4-like
There are no transcripts associated with this gene.
View Full Product Details