Product Details

SNP ID
rs4191803
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.16:59475735 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TCTGGGAACAAAAGCCCCTTCTCAT[C/T]CAGAGGGATGACCCTGTACTGGCCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
1700022E09Rik PubMed Links

Gene Details

Gene
1700022E09Rik
Gene Name
RIKEN cDNA 1700022E09 gene
There are no transcripts associated with this gene.

Gene
Mina
Gene Name
myc induced nuclear antigen
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025910.3 397 Silent Mutation NP_080186.3

View Full Product Details