Product Details
- SNP ID
-
rs13459167
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:19133719 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGAAAATGAGTTTTCCAATCACAT[A/G]CGCCTGTGAAACAGAATAAAGCTAC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Armc1
PubMed Links
Gene Details
- Gene
- Armc1
- Gene Name
- armadillo repeat containing 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_028840.2 |
1366 |
UTR 3 |
|
|
NP_083116.1 |
View Full Product Details