Product Details

SNP ID
rs13477595
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.4:20519098 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AGGAGTTGCAGTAGGGACTGTCTAG[A/T]GAAAGACTCTTAAATTTTATCTGAG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
Nkain3 PubMed Links

Gene Details

Gene
Nkain3
Gene Name
Na+/K+ transporting ATPase interacting 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172987.2 Intron NP_766575.2

View Full Product Details