Product Details
- SNP ID
-
rs6297540
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:103006064 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAAACCATTATTTAATTTGTTCTCA[G/T]CCCCAGCATCTTGTATGAAGTCCAA
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Dcakd
PubMed Links
Gene Details
- Gene
- Dcakd
- Gene Name
- dephospho-CoA kinase domain containing
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_026551.3 |
|
Intron |
|
|
NP_080827.2 |
- Gene
- Mir6931
- Gene Name
- microRNA 6931
There are no transcripts associated with this gene.
View Full Product Details