Product Details
- SNP ID
-
rs13481638
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:107974829 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTCTGAAATGTCACTGACTAAAGA[A/C]CGGAGCAGGCCTGTAATTACGTTCA
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Bcl11b
PubMed Links
Gene Details
- Gene
- Bcl11b
- Gene Name
- B cell leukemia/lymphoma 11B
View Full Product Details