Product Details

SNP ID
rs6200652
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.13:36972100 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GGATGGGAATACAGTGACCTTTAAC[C/T]AACCACTCGGATTCCAAACCCCTGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
F13a1 PubMed Links

Gene Details

Gene
F13a1
Gene Name
coagulation factor XIII, A1 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166391.1 Intron NP_001159863.1
NM_028784.3 Intron NP_083060.2
Gene
Gm30489
Gene Name
predicted gene, 30489
There are no transcripts associated with this gene.

View Full Product Details