Product Details

SNP ID
rs3089547
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:44456266 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AGTAGACTGAAAAAGTGGTTTCAGA[C/T]TTTTCCCACCTTATACAAAATTCAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Ublcp1 PubMed Links

Gene Details

Gene
Ublcp1
Gene Name
ubiquitin-like domain containing CTD phosphatase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024475.5 Intron NP_077795.2

View Full Product Details