Product Details

SNP ID
rs13481541
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.12:76378312 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AATTCAGAACTTCCCGTACTAACTA[C/T]TGAATTTAATAGCTAGCCAGAACGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Zbtb1 PubMed Links

Gene Details

Gene
Zbtb1
Gene Name
zinc finger and BTB domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178744.2 Intron NP_848859.1
Gene
Zbtb25
Gene Name
zinc finger and BTB domain containing 25
There are no transcripts associated with this gene.

View Full Product Details