Product Details

SNP ID
rs13483092
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.17:71001085 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AAAACTAGTAAATATATCCTTCCAA[A/T]TACTCCCATTTCAAAACCTCAGGTA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
Myl12a PubMed Links

Gene Details

Gene
Myl12a
Gene Name
myosin, light chain 12A, regulatory, non-sarcomeric
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_026064.2 Intron NP_080340.2

View Full Product Details