Product Details

SNP ID
rs13477380
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.3:123282622 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GAGGTGCCACTGGTGTCACTGCCAG[C/T]ACAGAGCCCTGGGTAGGGATGAGGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Sec24d PubMed Links

Gene Details

Gene
Sec24d
Gene Name
Sec24 related gene family, member D (S. cerevisiae)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_027135.2 Intron NP_081411.2

View Full Product Details