Product Details

SNP ID
rs13477936
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.4:117926857 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CGGAGCTGTGGCCTAGGCCGAGCGC[A/G]CTCACCGGCACCAGCTGCGAGCGCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Artn PubMed Links

Gene Details

Gene
Artn
Gene Name
artemin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284191.1 1630 Silent Mutation NP_001271120.1
NM_001284192.1 1630 Missense Mutation NP_001271121.1
NM_001284193.1 1630 Silent Mutation NP_001271122.1
NM_009711.4 1630 Silent Mutation NP_033841.1
Gene
St3gal3
Gene Name
ST3 beta-galactoside alpha-2,3-sialyltransferase 3
There are no transcripts associated with this gene.

View Full Product Details